missing translation for 'onlineSavingsMsg'
Learn More

Human IL-1Rrp2/IL-1R6 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£167.00 - £460.00

Specifications

Antigen IL-1Rrp2/IL-1R6
Clone 116004
Dilution Neutralization 0.25-1.5 ug/mL
Applications Neutralization
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18738948
GREENER_CHOICE
View Documents
R&D Systems
MAB8721-SP
25 μg
£167.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18734298
GREENER_CHOICE
View Documents
R&D Systems
MAB8721-100
100 μg
£460.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

IL-1Rrp2/IL-1R6 Monoclonal specifically detects IL-1Rrp2/IL-1R6 in Human samples. It is validated for Neutralization.
Specifications

Specifications

IL-1Rrp2/IL-1R6
Neutralization 0.25-1.5 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
IL-1 R6, IL-1 Rrp2, IL1RL2, IL-1RL2, IL1Rrp2, IL-1Rrp2, IL1RRP2IL1R-rp2Interleukin-1 receptor-related protein 2, interleukin 1 receptor-like 2, interleukin-1 receptor-like 2
Mouse myeloma cell line NS0-derived recombinant human IL-1 Rrp2/IL-1 R6 Asp20-Tyr337 Accession # Q9HB29
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human IL-1 Rrp2/IL-1 R6 in direct ELISAs.
116004
Neutralization
Unconjugated
Mouse
Human
Q9HB29
8808
IgG1
Protein A or G purified from ascites
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only (RUO)

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.