missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-18 R beta/IL-1 R7 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
£258.00 - £461.00
Specifications
| Antigen | IL-18 R beta/IL-1 R7/ACPL |
|---|---|
| Clone | 132029 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18786147
![]() |
R&D Systems
FAB118P-025 |
25 Tests |
£258.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18777966
![]() |
R&D Systems
FAB118P-100 |
100 Tests |
£461.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-18 R beta/IL-1 R7/ACPL Monoclonal specifically detects IL-18 R beta/IL-1 R7/ACPL in Human samples. It is validated for Flow Cytometry.Specifications
| IL-18 R beta/IL-1 R7/ACPL | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Accessory protein-like, AcPL, CD218 antigen-like family member B, CD218b, CD218b antigen, CDw218b, IL-1 R7, IL-18 receptor accessory protein, IL-18 receptor beta, IL18R beta, IL-18RAcP, IL18RAP, IL-18Rb, IL18RB, IL-18Rbeta, IL-18R-beta, IL-1R accessory protein-like, IL1R7, IL-1R7, IL-1R-7, IL-1RAcPL, interleukin 18 receptor accessory protein, Interleukin-1 receptor 7, interleukin-18 receptor accessory protein, Interleukin-18 receptor accessory protein-like, Interleukin-18 receptor beta, MGC120589, MGC120590 | |
| IL18RAP | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human IL-18 R beta/IL-1 R7 in direct ELISAs and Western blots. In direct ELISAs, 15% cross-reactivity with recombinant human (rh) IL-1 RII is observed and 2% cross-reactivity with rhIL-1 R9 is observed. Also in direct ELISAs, no cross-reactivity with rhIL-1 RI, rhIL-1 RacP/IL-1 R3, rhIL-1 R4, rhIL-18 R alpha/IL-1 R5, rhIL-1 R6, rhIL-1 R8, or recombinant mouse (rm) IL-18 R beta/IL-1 R7 is observed. In Western blots, approximately 100% cross-reactivity with rmIL-18 R beta is observed and no cross-reactivity with rhIL-18 R alpha/IL-1 R5, rhIL-1 RacP/IL-1 R3, rhIL-1 RI, rhIL-1 RII, or rhIL-1 Rrp2/IL-1 R6 is observed. |
| 132029 | |
| Flow Cytometry | |
| R-PE | |
| Mouse | |
| Human | |
| O95256 | |
| 8807 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-18 R beta/IL-1 R7 Met1-Arg356 Accession # O95256 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title