missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IL-18 R alpha/IL-1 R5 ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Description
Human IL-18 R alpha/IL-1 R5 quantitates human IL-18 R alpha/IL-1 R5 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-18 R alpha/IL-1 R5.
IL18R1 is a cytokine receptor that belongs to the interleukin 1 receptor family. This receptor specifically binds interleukin 18 (IL18), and is essential for IL18 mediated signal transduction. IFN-alpha and IL12 are reported to induce the expression of this receptor in NK and T cells. The gene along with four other members of the interleukin 1 receptor family, including IL1R2, IL1R1, ILRL2 (IL-1Rrp2), and IL1RL1 (T1/ST2), form a gene cluster on chromosome 2q.
Specifications
Specifications
| Accession Number | CD218 antigen-like family member A; CD218a; CDw218a; cytokine receptor; IL1 receptor-related protein; Il18r1; IL-18R1; IL-18 R-1; IL18RA; Il18ralpha; IL-18 R-alpha; IL18Ralpha2; Il1rrp; IL-1 Rrp; IL1R-rp; interleukin 1 receptor related protein; interleukin 18 receptor 1; Interleukin-18; interleukin-18 receptor 1; Interleukin-18 receptor alpha |
| Assay Range | 28.67 to 7,000 pg/mL |
| Assay Sensitivity | 28 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 8809 |
| Gene Symbol | Il18r1 |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction