missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-18 Human Instant ELISA™ Kit

Product Code. 15541857
Change view
Click to view available options
Quantity:
128 Tests
Unit Size:
128 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15541857 128 Tests 128 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15541857 Supplier Invitrogen™ Supplier No. BMS267INST

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

  • Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-18, HRP-Conjugate (human IL-18 monoclonal antibody) and Sample Diluent, lyophilized
  • Aluminum pouch(es) with a human IL-18 Standard curve (coloured)
  • Wash Buffer Concentrate 20x

Instant ELISA Kit

The Human Interleukin 18 (Hu IL-18) ELISA quantitates Hu IL-18 in human serum, plasma, cell culture supernatants, urine or other body fluids. The assay will exclusively recognize both natural and recombinant Hu IL-18. Principle of the method The Human IL-18 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14116
Assay Range 78-5,000 pg/mL
Assay Sensitivity 9.2 pg/mL
Conjugate Biotin
Product Type ELISA Kit
Sample Type Plasma, Serum, Supernatant, Urine
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 3606
Gene Symbol IGIF,IL-18,IL-1g,IL1F4
Interassay CV 8.1%
Intraassay CV 6.5%
Kit Contents Pre-coated 96 well Microplate, Biotinylated Detection Antibody, SAV-HRP, Sample Diluent, lyophilized, Standard curve strips, Wash Buffer Concentrate, Substrate Solution, Stop Solution, Lysis Buffer 10x, Adhesive Plate Covers
Label or Dye HRP
Quantity 128 Tests
Regulatory Status RUO
Sample Volume Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL; Urine,50 μL
Storage Requirements -20°C
Synonym Interleukin-18, Interferon-gamma-inducing factor
Target Human
Test Time 20 min.
Total Assay Time 3 hr. 10 min.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.