missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-18 BPa Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB119-SP
This item is not returnable.
View return policy
Description
IL-18 BPa/IL18BP Monoclonal specifically detects IL-18 BPa/IL18BP in Human samples. It is validated for Western Blot.
Specifications
| IL-18 BPa/IL18BP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| IL18 BPa, IL18BP, IL-18BP, IL18BPa, interleukin 18 binding protein, interleukin-18-binding protein, MC51L-53L-54L homolog gene product, tadekinig-alfa | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10068 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 136033 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IL18BP | |
| Mouse myeloma cell line NS0-derived recombinant human IL-18 BPa | |
| 25 μg | |
| Primary | |
| Detects human IL-18 BPa in direct ELISAs and Western blots. In direct ELISAs and Western blots, does not cross-react with recombinant mouse (rm) IL-18 BPc or rmIL-18 BPd. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction