missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-17RB Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB12071
This item is not returnable.
View return policy
Description
IL-17RB Monoclonal specifically detects IL-17RB in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| IL-17RB | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q9NRM6 | |
| IL17RB | |
| Mouse myeloma cell line NS0-derived recombinant human IL-17 RB Arg18-Gly289 Accession # Q9NRM6 | |
| 500 μg | |
| Primary | |
| Detects human IL-17 RB in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) IL-17 RB, recombinant human IL-17 R or rmIL-17 R is observed. | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 170216 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CRL4IL-17 receptor homolog 1, cytokine receptor CRL4, Cytokine receptor-like 4, Evi27, EVI27IL-17B receptor, IL-17 RB, IL-17 receptor B, IL-17 RH1, IL17BRinterleukin 17B receptor, IL-17ER, IL17RB, IL-17RB, IL17Rh1, IL-17Rh1, IL17RH1MGC5245, interleukin 17 receptor B, interleukin 17 receptor homolog 1, interleukin-17 receptor B, Interleukin-17B receptor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 55540 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction