missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-17RA/IL-17R (Research Grade Brodalumab Biosimilar) Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Human Monoclonal Antibody
Brand: R&D Systems FAB10547V-100UG
This item is not returnable.
View return policy
Description
IL-17RA/IL-17R Monoclonal antibody specifically detects IL-17RA/IL-17R in Human samples. It is validated for Flow Cytometry
Specifications
| IL-17RA/IL-17R | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Human | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 23765 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| Hu129 | |
| Flow Cytometry 0.25-1 ug/106 cells | |
| CD217, CD217 antigen, Cdw217, CDw217interleukin 17 receptor, hIL-17R, IL-17 R, IL-17 RA, IL-17 receptor A, IL17RA, IL-17RAMGC10262, IL17Rinterleukin-17 receptor A, interleukin 17 receptor A | |
| Human Il-17 Receptor | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction