missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-17F DuoSet ELISA
Click to view available options
Quantity:
1 Kit (for 15 Plates)
Unit Size:
1 set
Description
Includes
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
The Human IL-17F DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-17F.
Specifications
Specifications
| Quantity | 1 Kit (for 15 Plates) |
| Species | Human |
| Conjugate | Biotin |
| Binding Target | IL-17F |
| Assay Range | 12.50 to 800 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 112744 |
| Synonym | Cytokine ML-1, IL17F, IL-17Finterleukin-17F, IL24, IL-24, interleukin 17F, Interleukin-24, ML-1, ML1interleukin-24 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction