missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-17E DuoSet ELISA
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
1 set
Description
Includes
Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
The Human IL-17E DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-17E/IL-25.
Specifications
Specifications
| Description | IL17E, IL-17E, IL17Einterleukin-17E, IL25, IL-25, interleukin 17E, interleukin 25, Interleukin-17E, interleukin-25 |
| Product Type | ELISA Kit |
| Quantity | 1 Kit (for 5 Plates) |
| Species | Human |
| Cross Reactivity | Not observed |
| Format | 96-well Strip Plate |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 64806 |
| Sample Type | Cell Culture Supernatant, Serum, Plasma |
| Kit Contents | Capture antibody, Detection antibody, Recombinant standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction