missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-17C Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF1234
This item is not returnable.
View return policy
Description
IL-17C Polyclonal specifically detects IL-17C in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| IL-17C | |
| Polyclonal | |
| Biotin | |
| Q9P0M4 | |
| IL17C | |
| E. coli-derived recombinant human IL-17C His19-Val197 Accession # Q9P0M4 | |
| 50 μg | |
| Primary | |
| Detects human IL-17C in ELISAs and Western blots. In sandwich immunoassays, approximately 10% cross-reactivity with recombinant mouse (rm) IL-17C is observed and less than 0.3% cross-reactivity with recombinant human (rh) IL-17, rhIL-17B, rhIL-17D, rhIL-17E, and rhIL-17F is observed. | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| CX2, Cytokine CX2, IL17C, IL-17CMGC126884, interleukin 17C, interleukin-17C, MGC138401 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 27189 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction