missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-17B Antibody, R&D Systems™

Mouse Monoclonal Antibody
£170.00 - £1525.00
Specifications
| Antigen | IL-17B |
|---|---|
| Clone | 174113 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Flow Cytometry, CyTOF, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18780587
![]() |
R&D Systems
MAB1248-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18777709
![]() |
R&D Systems
MAB1248-100 |
100 μg |
£379.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18711917
![]() |
R&D Systems
MAB1248-500 |
500 μg |
£1525.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-17B Monoclonal specifically detects IL-17B in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
| IL-17B | |
| LYOPH | |
| Western Blot, Flow Cytometry, CyTOF, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL17B, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20 | |
| IL17B | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 174113 | |
| Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q9UHF5 | |
| 27190 | |
| E. coli-derived recombinant human IL-17B Gln21-Phe180 Accession # Q9UHF5 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-17B in Western blots and ELISAs. In Western blots, approximately 25% cross-reactivity with recombinant mouse IL-17B is observed and no cross-reactivity with recombinant human (rh) IL-17, rhIL-17C, rhIL-17E or rhIL-17F is observed. In sandwich immunoassays, less than 0.5% cross-reactivity with rmIL-17B, and no cross-reactivity with rhIL-17C, rhIL-17D, rhIL-17E, rhIL-17F, or rhIL-17 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title