missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-17AF Human ELISA Kit

For the quantitative detection of human IL-17AF.

Brand:  Invitrogen™ BMS2082TEN

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15511607

  • £3070.00 / Pack of 10
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-17A
Biotin-Conjugate anti-human IL-17F monoclonal antibody
Streptavidin-HRP
Human IL-17AF Standard lyophilized, 2000pg/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Calibrator Diluent
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

The Human Interleukin-17AF (Hu IL-17AF) ELISA quantitates Hu IL-17AF in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-17AF. Principle of the method The Human IL-17AF solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Interleukin-17A (IL-17A) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. The cDNA encoding human IL-17A was isolated from a library of CD4+ T cells; the encoded protein exhibits 72 percent amino acid identity with HVS13 , an open reading frame from a T lymphotropic Herpesvirus saimiri, and 63 percent with mouse CTLA-8 (cytotoxic T-lymphocyte associated antigen-8). Human IL-17A exists as glycosylated 20-30 kD homodimers. High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells. IL-17A enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. Human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A binds to a receptor that binds also to HVS13 (viral IL-17) and to CTLA-8.This recombinant protein is E. coli expressed. Insect cell expressed Human IL-17A Recombinant Protein (cat. 14-8180) is also available.
Specifications

Specifications

Q16552,Q96PD4
8.8 pg/mL
ELISA Kit
Human
Colorimetric Microplate Reader
CANDF6,IL-17F,ML-1,ML1,Ctla-8,Ctla8,IL-17,IL-17A,Il17
5.9%
HRP
RUO
2°C to 8°C
Human
3 hr. 30 min.
31.3-2,000 pg/mL
Biotin
Plasma, Serum, Supernatant
ELISA
112744, 3605
5.2%
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
10 x 96 Tests
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL
Interleukin 17AF, IL-17 A/F heterodimer
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only