missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IL-17 RB ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
Description
Human IL-17 RB quantitates human IL-17 RB in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-17 RB.
Recently, a number of cytokines belonging to the interleukin (IL)-17 family have been identified. These are termed as IL-17B, IL-17C (1) and IL-17E. IL-17 is a potent proinflammatory cytokine that plays roles in a number of diseases including rheumatoid arthritis, multiple sclerosis, and promotion of tumor growth. IL-17B, C, and E like IL-17 are able to induce proinflammatory responses. However, they do not bind to the IL-17 receptor suggesting that additional IL-17R related receptor might exist. Receptor for IL-17B and IL-17E has been independently isolated and has been designated as EV127 (in mouse) and IL-17Rh1 (in human), respectively. IL-17E induces activation of NF-kB pathway and like IL-17 also induces production of IL-8. IL-17Rh1 mRNA is highly expressed in Kidney and liver and at moderate level in small intestine, colon, and brain.
Specifications
Specifications
| Accession Number | CRL4; cytokine receptor CRL4; cytokine receptor-like 4; ecotropic viral integration site 27; Evi27; IL-17 receptor B; IL-17 receptor homolog 1; IL-17 B; IL-17 B receptor; IL17BR; IL-17 ER; IL17RB; IL-17 RB; IL17RH1; IL-17 Rh1; IL-20; interleukin 17 receptor B; interleukin 17 receptor homolog 1; interleukin 17 B receptor; Interleukin-17 receptor B; interleukin-17 B receptor; MGC138900; MGC138901; MGC5245; UNQ2501/PRO19612; ZCYTO7 |
| Assay Range | 0.85 to 200 ng/mL |
| Assay Sensitivity | 0.85 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 55540 |
| Gene Symbol | IL17RB |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction