missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-17/IL-17A PerCP-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems IC3171C-100
This item is not returnable.
View return policy
Description
IL-17/IL-17A Monoclonal specifically detects IL-17/IL-17A in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-17/IL-17A | |
| Monoclonal | |
| PerCP | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3605 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 41802 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| Q16552 | |
| IL17A | |
| E. coli-derived recombinant human IL-17 Ile20-Ala155 Accession # Q16552 | |
| 100 Tests | |
| Primary | |
| Detects human IL-17 in direct ELISAs. In direct ELISAs, approximately 40% cross-reactivity with recombinant human (rh) IL-17A/IL-17F heterodimer is observed. No cross-reactivity with recombinant mouse IL-17, recombinant canine Il-17, rhIL-17B, rhIL-17C, rhIL-17D, rhIL-17E, or rhIL-17F is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction