missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-15 Recombinant Protein

Product Code. p-5232740
Click to view available options
Quantity:
1 mg
10 μg
100 μg
Unit Size:
1mg
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10013534

Brand: Gibco™ PHC9151

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

Carrier-Free Reconstitution: We recommend reconstituting the lyophilized protein in sterile double distilled water containing at least 0.1% human serum albumin or bovine serum albumin to prepare a stock solution of =1 mg/mL of the protein. The reconstituted protein should be apportioned into working aliquots and stored at =-20°C. Storage: The lyophilized protein is stable for a few weeks at room temperature, but is best stored at -20°C. Reconstituted human IL-15 should be stored at =-20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.

IL-15 (Interleukin 15 ) is a cytokine that regulates T and natural killer cell activation and proliferation. IL-15 and IL-2 share many biological activities as both have been found to bind common hematopoietin receptor subunits, and may compete for the same receptor, and thus negatively regulate each other's activity. The number of CD8+ memory cells is shown to be controlled by a balance between IL-15 and IL-2. IL-15 induces the activation of JAK kinases, as well as the phosphorylation and activation of transcription activators STAT3, STAT5, and STAT6. In mouse, studies suggest that IL-15 may increase the expression of apoptosis inhibitor Bcl-xL, possibly through the transcription activation activity of STAT6, and thus prevent apoptosis. IL-15 plays an important role in the growth and differentiation of T and B lymphocytes, natural killer cells, macrophages, and monocytes as well as activation of a number of important intracellular signaling molecules. As such, IL-15 could be essential for the immune responses, allograft rejection, and the pathogenesis of autoimmune diseases. Further, IL-15 is a widely expressed pro-inflammatory cytokine and has been shown to play a role in several inflammatory disorders, including rheumatoid arthritis, psoriasis and pulmonary inflammatory diseases. Emerging data suggest that IL-15 may serve as a good therapeutic target, as there appears to be a beneficial effect of IL-15 neutralization in models of psoriasis and diabetes.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P40933
For Use With (Application) Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 3600
Molecular Weight (g/mol) 12.9 kDa
Name Human IL-15
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/mL
Gene Alias AI503618; il 15; Il15; IL-15; ILN; Interleukin; interleukin 15; Interleukin15; interleukin-15; MGC9721; M-IL-15
Common Name IL-15
Gene Symbol IL15
Biological Activity ED50 ≤ 0.5 ng/mL; determined by the dose-dependent proliferation of CTLL-2 cells.
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sequence Human IL-15 recombinant protein contains 115 amino acids
Content And Storage -20°C
Classification Carrier-Free
Activity ED50 ≤ 0.5 ng/mL; determined by the dose-dependent proliferation of CTLL-2 cells.
Endotoxin Level <1 EU/ml
Shipping Condition Approved for shipment on Wet or Dry Ice
Expression System E. coli
Protein Family Cytokines & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype Interleukins
Research Category Cell Cycle & Proliferation, Oncology, Immunology, Signal Transduction
Product Line Gibco
Purity or Quality Grade ≥98% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.