missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-15 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£201.00 - £522.00
Specifications
| Antigen | IL-15 |
|---|---|
| Clone | 1048222 |
| Dilution | Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 3-25 ug/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18722227
![]() |
R&D Systems
MAB2472-SP |
25 μg |
£201.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18793477
![]() |
R&D Systems
MAB2472-100 |
100 μg |
£522.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-15 Monoclonal antibody specifically detects IL-15 in Human samples. It is validated for Immunohistochemistry, ImmunocytochemistrySpecifications
| IL-15 | |
| Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 3-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| IL15, IL-15MGC9721, interleukin 15, interleukin-15 | |
| E. coli-derived human IL-15 protein, Asn49-Ser162, Accession # P40933.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1048222 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P40933.1 | |
| 3600 | |
| IgG2a | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title