Learn More
Gibco™ Human IL-13 Recombinant Protein
Recombinant Human Interleukin 13 (IL13) is a bioactive protein intended for use in cell culture applications
Brand: Gibco™ PHC0135
Code : 50
Additional Details : Weight : 0.50000kg
Description
Human IL-13 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL13 down-regulates macrophage produced cytokines such as TNF-α, IL-1, IL-6, IL-8, and IL-12 in response to LPS challenge or IFN-γ. IL13 induces monocyte differentiation and also induces B cell differentiation and proliferation.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human TF-1 cells
- Proven compatibility: Gibco proteins are bioassayed with Gibco media
Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
17 kDa | |
25μg | |
Human | |
Carrier-Free | |
25 μg | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
3596 | |
Sequential Chromatography | |
IL-13 | |
Store in freezer (-5 to -30°C). | |
< 0.1 ng/μg | |
Wet Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Respiratory Research, Inflammation, Signal Transduction | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.