Learn More
Gibco™ Human IL-12 Recombinant Protein
Description
Carrier-Free Reconstitution: We recommend that the vial be briefly centrifuged prior to opening to bring the contents to the bottom. This lyophilized protein should be reconstituted in phosphate buffered saline, pH 7.2-pH 7.4, to a concentration of ≥0.2 mg/mL. Further dilutions should be made in sterile medium or buffered solution with a carrier protein. Storage: This lyophilized preparation is stable at 2°C to 8°C, but should be kept at -20°C for long term storage, preferably desiccated. Upon reconstitution, the preparation is stable for up to one week at 2°C to 8°C. For maximal stability, apportion the reconstituted preparation into working aliquots and store at ≤-20°C. Avoid repeated freeze/thaw cycles.
Specifications
Specifications
| Accession Number | P29459, P29460 |
| For Use With (Application) | Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 3592, 3593 |
| Molecular Weight (g/mol) | 75 kDa |
| Name | Human IL-12 |
| pH Range | 7.4 |
| Purification Method | Purified |
| Quantity | 10 μg |
| Storage Requirements | -20°C |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.