missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-12 R beta 2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF1959
Additional Details : Weight : 0.00970kg
Description
IL-12 R beta 2 Polyclonal specifically detects IL-12 R beta 2 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Specifications
IL-12 R beta 2 | |
Polyclonal | |
Unconjugated | |
Q99665 | |
IL12RB2 | |
Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 2 Cys28-Asn622 Accession # Q99665 | |
100 μg | |
Primary | |
Detects human and mouse IL-12 R beta 2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human IL-12 R beta 1 is observed. | |
Human | |
IgG |
Western Blot, Blockade of Receptor-ligand Interaction | |
LYOPH | |
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
IL-12 receptor beta 2, IL-12 receptor subunit beta-2, IL12R beta 2, IL-12R subunit beta-2, IL12RB2, IL-12RB2, IL-12R-beta-2, interleukin 12 receptor, beta 2, interleukin-12 receptor beta-2 chain, interleukin-12 receptor subunit beta-2 | |
Goat | |
Affinity Purified | |
RUO | |
3595 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only