missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-12 p70 Recombinant Protein, PeproTech®

Product Code. 17861293
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17861293 100 μg 100µg
17831093 10 μg 10µg
17860823 1 mg 1mg
17840223 250 μg 250µg
17801083 2 μg 2µg
17850903 500 μg 500µg
17811303 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 17861293 Supplier Gibco™ Supplier No. 20012H100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

PeproTech's Recombinant Human IL-12 p70, derived from HEK293 cells, is a heterodimeric glycoprotein consisting of disulfide-linked p35 and p40 subunits (503 total amino acid residues). The calculated molecular weight of Human IL-12 p70 is 57.2 kDa; however, due to glycosylation, it migrates at an apparent molecular weight of 65-70 kDa based on SDS-PAGE gel, under non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

IL-12 p70, also known as interleukin-12, is a cytokine that plays a critical role in the immune system's response to infections and cancer. It is produced by immune cells, including dendritic cells, macrophages, and B cells, in response to certain pathogens and tumor cells. IL-12 p70 stimulates the production of interferon-gamma and other cytokines, which promote the activation and proliferation of T cells and natural killer cells. IL-12 p70 has been studied as a potential therapeutic target for various diseases and conditions, including infectious diseases, autoimmune disorders, and cancer. It has been shown to enhance immune responses against certain infections and tumors, and to modulate inflammatory responses in autoimmune diseases.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P29459, P29460
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 3592, 3593
Molecular Weight (g/mol) 75 kDa
Name Human IL-12 p70
Quantity 100 μg
Source HEK293 cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 20C2; CLMF; CLMF p35; CLMF p40; CLMF2; cytokine; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; Cytotoxic lymphocyte maturation factor 40 kDa subunit; il 12; IL 12p70; Il12; IL-12; Il-12 p35; IL-12 p75; IL-12 subunit p35; IL-12 subunit p40; IL-12, subunit p35; IL12, subunit p40; Il-12; cytokine; IL12A; IL-12 A; IL12A/B; IL12AB; IL12B; Il-12 b; IL12P35; IL-12p35; Il12p40; Il-12p40; IL35 subunit; ILN; IMD28; IMD29; Interleukin; interleukin 12 35 kDa subunit; interleukin 12 35 kDa subunit; interleukin 12 40 kDa subunit; interleukin 12 40 kDa subunit; interleukin 12 p35 subunit; interleukin 12 p40 subunit precursor; interleukin 12 polypeptide 35; interleukin 12, p35; interleukin 12, p40; interleukin 12 A; interleukin 12 A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin 12 B; interleukin 12 B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleukin 12p35 subunit precursor; interleukin-12 alpha chain; interleukin-12 alpha p40 subunit; interleukin-12 beta chain; interleukin-12 beta p35 subunit; interleukin-12 p35 subunit; interleukin-12 p35 subunit precursor; interleukin-12 p40; interleukin-12 p40 subunit; interleukin-12 p40 subunit precursor; Interleukin-12 p70; interleukin-12 subunit alpha; Interleukin-12 subunit beta; interleukin-23 p40 subunit; Ll12a; MGC151228; MGC151232; natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35; natural killer cell stimulatory factor, 40 kD subunit; NF cell stimulatory factor chain 1; NFSK; NK cell stimulatory factor chain 1; NK cell stimulatory factor chain 2; NKSF; NKSF1; NKSF2; P35; p40
Common Name IL-12 p70
Gene Symbol IL12A, Il12b
Biological Activity Determined by its ability to increase IFN-gamma production by anti-TCR mAb-stimulated PBMCs.
Conjugate Unconjugated
Recombinant Recombinant
Sequence p35 subunit: RNLPVATPDP GMFPCLHHSQ NLLRAVSNML QKARQTLEFY PCTSEEIDHE DITKDKTSTV EACLPLELTK NESCLNSRET SFITNGSCLA SRKTSFMMAL CLSSIYEDLK MYQVEFKTMN AKLLMDPKRQ IFLDQNMLAV IDELMQALNF NSETVPQKSS LEEPDFYKTK IKLCILLHAF RIRAVTIDRV MSYLNAS p40 subunit: IWELKKDVYV VELDWYPDAP GEMVVLTCDT PEEDGITWTL DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSHSLL LLHKKEDGIW STDILKDQKE PKNKTFLRCE AKNYSGRFTC WWLTTISTDL TFSVKSSRGS SDPQGVTCGA ATLSAERVRG DNKEYEYSVE CQEDSACPAA EESLPIEVMV DAVHKLKYEN YTSSFFIRDI IKPDPPKNLQ LKPLKNSRQV EVSWEYPDTW STPHSYFSLT FCVQVQGKSK REKKDRVFTD KTSATVICRK NASISVRAQD RYYSSSWSEW ASVPCS
Content And Storage -20°C
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.