Learn More
Invitrogen™ Human IL-12/IL-23 p40 ELISA Kit
Beskrivning
Innehåller
Aluminium pouch(es) with a Microwell Plate coated with polyclonal antibody to human IL-12/IL-23 p40
Biotin-Conjugate anti-human IL-12/IL-23 p40 monoclonal antibody
Streptavidin-HRP
Human IL-12/IL-23 p40 Standard lyophilized, 4ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
The Human Interleukin-12/Interleukin-23 p40 subunit (Hu IL-12/IL-23 p40) ELISA quantitates Hu IL-12/IL-23 p40 in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-12/IL-23 p40. Principle of the method The Human IL-12/IL-23 p40 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifikationer
Specifikationer
| Accession Number | P29459, P29460, Q9NPF7 |
| Assay Range | 31.3-2000 pg/mL |
| Assay Sensitivity | <10 pg/mL |
| Content And Storage | Aluminum pouch(es) with a Microplate coated with polyclonal antibody to human IL-12/IL-23 p40 Biotin-Conjugate anti-human IL-12/IL-23 p40 monoclonal antibody Streptavidin-HRP Human IL-12/IL-23 p40 Standard lyophilized, 4 ng/mL upon reconstitution Sample Diluent Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA) Wash Buffer Concentrate 20x (PBS with 1% Tween 20) Substrate Solution (tetramethyl-benzidine) Stop Solution (1M Phosphoric acid) Adhesive Films |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Visa mer |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.