Learn More
Human IL-11 R alpha Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 2 publications
Brand:  R&D Systems MAB1977
 View more versions of this product
Product Code. 18139240
  • £422.00 / 100µg
Description
IL-11R alpha Monoclonal specifically detects IL-11R alpha in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-11R alpha
Monoclonal
LYOPH
Western Blot 1 ug/mL
Q14626
IL11RA
Mouse myeloma cell line NS0-derived recombinant human IL-11 R alpha Cys26-Val363 Accession # Q14626
100 μg
Primary
Detects human IL-11 R alpha in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with rmIL-11 R alpha or recombinant human IL-6 R alpha is observed. In Western blots, 25% cross-reactivity with recombinant mouse (rm) IL-11 R alpha is observed.
Human
Purified
Western Blot
473143
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
AI314697, GP130, IL11R alpha, IL11RA, IL-11RA, Il11ra2, NR1
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3590
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1