missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-11 Monoclonal Antibody (12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA530696
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human IL-11/interleukin 11.
Interleukins (ILs) are a large group of cytokines that are produced mainly by leukocytes, although some are produced by certain phagocytes and auxiliary cells. ILs have a variety of functions, but most function to direct other immune cells to divide and differentiate. Each IL acts on a specific, limited group of cells through a receptor specific for that IL.
Specifications
| IL-11 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P20809 | |
| IL11 | |
| Recombinant Human IL-11/interleukin 11 Protein (Pro22-Leu199). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| 12 | |
| Unconjugated | |
| IL11 | |
| adipogenesis inhibitory factor; AGIF; il 11; IL10RA; Il11; IL-11; IL11 inhibitor; IL-11ra; ILN; Interleukin; interleukin 11; Interleukin11; interleukin-11; MGC2146; Oprelvekin | |
| Mouse | |
| Protein A | |
| RUO | |
| 3589 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction