missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-10 Recombinant Protein

Product Code. 11528941
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11528941 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11528941 Supplier Invitrogen™ Supplier No. RP75613

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This product contains purified recombinant human IL-10 protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.

IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P22301
Concentration 0.505 mg/mL
For Use With (Application) Control, Western Blot
Formulation 0.01 M tris HCl with 50% glycerol and no preservative; pH 8
Gene ID (Entrez) 3586
Name Human IL-10
pH Range 8
Quantity 1 mg
Source E. coli
Immunogen N/A
Regulatory Status RUO
Gene Alias CSIF; cytokine; Cytokine synthesis inhibitory factor; GVHDS; H-IL-10; il 10; IL10; IL-10; IL-10 precursor; IL10A; IL10X; ILN; Interleukin; interleukin 10; Interleukin10; interleukin-10; MGC126450; MGC126451; RP11-262N9.1; T-cell growth inhibitory factor; TGIF
Common Name IL-10
Gene Symbol IL10
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Ser19- Asn178 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.