missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-10 Quantikine ELISA Kit
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 Plate
Unit Size:
1 set
Description
The Human IL-10 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-10 in cell culture supernates (200 ul), serum (200 ul), edta plasma (200 ul), heparin plasma (200 ul), citrate plasma (200 ul).
Specifications
Specifications
| Assay Range | 7.80 to 500 pg/mL |
| Assay Sensitivity | 3.9 pg/mL |
| Conjugate | HRP |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Citrate, Plasma (EDTA, Citrate, Heparin), Serum |
| Specificity | Natural and Recombinant Human IL-10 |
| Sufficient For | 1 x 96-well Plate |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction