Learn More
Human IL-1 RI PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems FAB269P-025
 View more versions of this product
Product Code. 18797657
  • £260.00 / 25 tests
Description
IL-1 RI Polyclonal specifically detects IL-1 RI in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-1 RI
Polyclonal
Flow Cytometry 10 uL/10^6 cells
CD121 antigen-like family member A, CD121a, CD121a antigen, IL1R, IL1R1, IL1RI, IL-1RI, IL1RT1, IL-1RT1, IL-1RT-1, Interleukin 1 Receptor 1, interleukin 1 receptor, type I, interleukin receptor 1, interleukin-1 receptor type 1, p80
Goat
Affinity Purified
RUO
3554
Human
IgG
Flow Cytometry
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
IL1R1
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human IL-1 RI
25 Tests
Primary
Detects human IL-1 RI in direct ELISAs and Western blots.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.