Learn More
Gibco™ Human IL-1 beta Recombinant Protein
Recombinant Human Interleukin 1, beta (IL1B) is a bioactive protein intended for use in cell culture applications
Brand: Gibco™ PHC0816
Code : 50
Additional Details : Weight : 0.50000kg
Description
Human IL-1 beta Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL-1 is involved in a number of biological processes including the stimulation of T-helper cells, promotion proliferation and synthesis of immunoglobulins, and the activation of fibroblasts and NK cells.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of murine D10S cells
- Proven Compatibility: Gibco proteins are bioassayed with Gibco media
Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
17.5 kDa | |
25μg | |
Human | |
Carrier-Free | |
25 μg | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
3553 | |
Sequential Chromatography | |
IL-1β | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
Wet Ice | |
Cytokines & Receptors | |
Lyophilized | |
Stem Cell Research, Immunology | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.