missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-1 beta ELISA Kits

Product Code. 10713295
Click to view available options
Quantity:
1 Plate Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10713295

Brand: Invitrogen™ EH2IL1B

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Measure levels of human interleukin-1 beta (IL-1b) in human serum, plasma (EDTA, heparin, sodium citrate), urine and culture supernatant samples using 96-well plates.

Assay IL-1b (human interleukin-1 beta) levels in human serum, plasma (EDTA, heparin, sodium citrate), urine and culture supernatant samples with our Invitrgen™ Human IL-1 beta ELISA Kits.

These human IL-1 beta ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-1 beta (IL-1β) in samples at 50 to 100μL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. The economically-priced reagent kit provides validated reagents only, requiring users to coat their own plates. The antibody-pair kit consists of the matched IL-1β antibody pairs and IL-1β standard only. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant human interleukin-1 beta (IL-1β)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 10.2 to 400pg/mL

  • Sensitivity: ≤ 1pg/mL

  • Precision: Intra-assay CV ≤ 10%; inter-assay CV ≤ 10%

  • Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 4 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with human IL-1α, IL-1RA, IL-2, IL-3, IL-4, IL-6, IL-7, IL-8, TNFα, IFNα, IFNγ or mouse IL-1β

Assay range and other values listed above are for the pre-coated plate kit; see product documentation for values specific to the reagent kit and antibody-pair kit.

Includes:Complete Kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates. The Reagent Kit (previously called a “Screening Set”) contains matched antibodies, standard and detection reagents but no coated plates. The Antibody Pair Kit (previously called a “MiniKit”) contains only matched antibodies and standard (no plates or detection reagents).

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 10.2 to 400pg/mL
Assay Sensitivity ≤ 1pg/mL
Description Human IL-1 beta ELISA Kit
Immunoassay Kit Format Colorimetric ELISA (TMB substrate) with Plates
Sample Type Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Specificity No significant cross-reaction with human IL-1a, IL-1RA, IL-2, IL-3, IL-4, IL-6, IL-7, IL-8, TNFa, IFNa, IFNg or mouse IL-1ß
Sufficient For 96 microplate wells
Kit Contents HRP Dilution Buffer, 14mL, 30X Wash Buffer, 50mL, Adhesive Plate Covers, 6 ea, Biotinylated Antibody Reagent, 8mL, Anti-Human IL-1 beta Precoated 96-well Strip Plate, 1 ea, Lyophilized Recombinant Human IL-1 beta Standard, 2 vials, TMB Substrate Solution, 13mL, Streptavidin-HRP Concentrate, 75 μL, Stop Solution, 13mL, Standard Diluent, 12mL
Measurement Absorbance at 450nm minus absorbance at 550nm
Precision Intra-assay CV ≤ 10%; inter-assay CV ≤ 10%
Quantity 1 Plate Kit
Sample Volume 50 μL
Target Natural and recombinant human interleukin-1 beta (IL-1ß)
Total Assay Time 4 hr.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.