Learn More
Gibco™ Human IL-1 alpha Recombinant Protein
Recombinant IL1A is a bioactive protein intended for use in cell culture applications
Brand: Gibco™ PHC0013
Code : 50
Additional Details : Weight : 0.50000kg
Description
Human IL-1 alpha Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL-1 is involved in a number of biological processes including the stimulation of T-helper cells, promotion proliferation and synthesis of immunoglobulins, and the activation of fibroblasts and NK cells.Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Cell Culture | |
Membrane-Filtered | |
IL-1α | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Cell Cycle & Proliferation, Immunology, Angiogenesis | |
95 % |
18.2 kDa | |
1mg | |
Human | |
Carrier-Free | |
1 mg | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.