missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-1 alpha Recombinant Protein

Product Code. 10534424
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
5 μg
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10534424 100 μg Each
10735593 1 mg Each
10405343 5 μg Each
10632084 25 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10534424 Supplier Gibco™ Supplier No. PHC0011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

Carrier-Free Reconstitution: We recommend that the vial be briefly centrifuged prior to opening to bring the contents to the bottom. Lyophilized human IL-1 alpha should be reconstituted in deionized water to 0.1-1.0 mg/mL to regain full activity. These stock solutions should be apportioned into working aliquots and stored at ≤-20°C. Further dilutions should be made in low endotoxin medium or buffered solution with heat inactivated FCS or tissue culture grade BSA. Storage: Lyophilized human IL-1 alpha should be stored at 2°C to 8°C, preferably desiccated. Store reconstituted human IL-1 alpha at ≤-20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.

IL-1 alpha (Interleukin-1 alpha) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 alpha is coded by the IL1A gene and signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 beta. IL1A is located on the q arm on chromosome 2 at position 13. IL-1 alpha plays an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses. IL-1 alpha activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 alpha is a pleiotropic cytokine involved in various immune responses, inflammatory processes, and hematopoiesis. Further, IL-1 alpha is produced by monocytes and macrophages as a proprotein, which is proteolytically processed and released in response to cell injury and induces apoptosis. The IL1A gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. IL1 alpha regulates the activities of NF-kappaB and mitogen-activated protein kinases. IL-1 alpha can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO). High levels of IL-1 alpha are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01583
For Use With (Application) Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 3552
Molecular Weight (g/mol) 18 kDa
Name Human IL-1 alpha
Purification Method Purified
Quantity 100 μg
Storage Requirements 4°C
Regulatory Status RUO
Endotoxin Concentration <0.1 ng/μg
Gene Alias CTLA8; hematopoietin-1; IL 1 a; IL 1α; IL1; IL-1 alpha; IL17; IL1A; Il-1a; IL-1alpha; IL1-ALPHA; IL1F1; IL1α; ILN; Interleukin; interleukin 1 alpha; interleukin 1, alpha; interleukin 17; interleukin 1a; interleukin 1-alpha; interleukin 1-alpha (AA 1 - 152); Interleukin1 alpha; interleukin-1 alpha; interleukin-1 alpha precursor; Interleukin-17A; preinterleukin 1 alpha; pre-interleukin-1 alpha; pro-interleukin-1-alpha; RP23-160G19.8
Common Name IL-1 alpha
Gene Symbol Il1a
Biological Activity ED50 = 0.001 - 0.005 ng/mL; determined by the dose-dependent proliferation of mouse D10S cells.
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sequence SAPFSFLSNV KYNFMRIIKY EFILNDALNQ SIIRANDQYL TAAALHNLDE AVKFDMGAYK SSKDDAKITV ILRISKTQLY VTAQDEDQPV LLKEMPEIPK TITGSETNLL FFWETHGTKN YFTSVAHPNL FIATKQDYWV CLAGGPPSIT DFQILENQA
Content And Storage 4°C
Classification Carrier-Free
Activity ED50 = 0.001 - 0.005 ng/mL; determined by the dose-dependent proliferation of mouse D10S cells.
Endotoxin Level <0.1 ng/μg
Shipping Condition Approved for shipment on Wet or Dry Ice
Expression System E. coli
Protein Family Cytokines & Receptors
Protein Form Recombinant, Ligand
Form Lyophilized
Protein Subtype Interleukins
Research Category Cell Cycle & Proliferation, Immunology, Angiogenesis
Product Line Gibco
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.