missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-1 alpha/IL-1F1 Membrane Form Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2001-500
This item is not returnable.
View return policy
Description
IL-1 alpha/IL-1F1 Monoclonal specifically detects IL-1 alpha/IL-1F1 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-ready.
Specifications
| IL-1 alpha/IL-1F1 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, Neutralization 5-20 ug/mL, CyTOF-ready | |
| P01583 | |
| IL1A | |
| E. coli-derived recombinant human IL-1 alpha/IL-1F1 Ser113-Ala271 Accession # P01583 | |
| 500 μg | |
| Primary | |
| Detects human IL-1 alpha/IL-1F1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IL-1 alpha , recombinant rat IL-1 alpha , recombinant cotton rat IL-1 alpha , recombinant human (rh) IL-1 beta or rhIL-18 is observed. | |
| Human | |
| Ascites |
| Flow Cytometry, Neutralization, CyTOF | |
| 3405 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BAF, Hematopoietin-1, IL1, IL1 alpha, IL-1 alpha, IL1A, IL-1A, IL1-ALPHA, IL1F1, IL-1F1, IL1F1hematopoietin-1, interleukin 1, alpha, interleukin-1 alpha, LAF, LEM, preinterleukin 1 alpha, pro-interleukin-1-alpha | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3552 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction