missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IGSF4C/SynCAM4 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB4164S-100UG
This item is not returnable.
View return policy
Description
IGSF4C/SynCAM4 Monoclonal specifically detects IGSF4C/SynCAM4 in Human samples. It is validated for Flow Cytometry.
Specifications
| IGSF4C/SynCAM4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8NFZ8 | |
| CADM4 | |
| Mouse myeloma cell line NS0-derived recombinant human IGSF4C/SynCAM4 Gln25-Tyr323 Accession # Q8NFZ8 | |
| RUO | |
| 199731 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 404619 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CADM4, cell adhesion molecule 4, IgSF4C, IGSF4Cnectin-like 4, Immunoglobulin superfamily member 4C, immunoglobulin superfamily, member 4C, Necl-4, NECL4synCAM4, Nectin-like protein 4, SYNCAM4, TSLC1-like 2, TSLC1-like protein 2, TSLL2, TSLL2SynCAM4 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects recombinant human (rh) IGSF4C/SynCAM4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) IGSF4A, rhIGSF4B, rhIGSF4D, or rhIGSF8 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction