missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IgM Human ELISA Standard (for Uncoated ELISA Kit)
Brand: Invitrogen™ 39-50620-65
Code : NEW
Additional Details : Weight : 0.47500kg
Description
Component of Human IgM ELISA Ready-SET-Go! KitSpecifications
Kit | |
Recombinant | |
Ready-SET-Go!, eBioscience | |
Human | |
Human | |
Liquid |
4 Pack | |
Microplate Reader | |
ELISA Standard | |
IgM | |
Human |
For Research Use Only. Not for use in diagnostic procedures.