missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IgM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA586030
This item is not returnable.
View return policy
Description
Human IgM Polyclonal Antibody for Western Blot, IHC (P)
IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
Specifications
| Human IgM | |
| Polyclonal | |
| Unconjugated | |
| IGHM | |
| AGM1; IGHM; immunoglobulin heavy constant mu; MU; VH | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3507 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| P01871 | |
| IGHM | |
| Human IgM from Human serum. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction