missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IgM Human Uncoated ELISA Kit with Plates

For quantitative detection of human IgM.

Brand:  Invitrogen™ 88-50620-86

Code : NEW

Additional Details : Weight : 0.47500kg

 View more versions of this product

Product Code. 15590957

  • £392.00 / Pack of 10
Estimated Shipment: 02-05-2024
to see stock.
Add to basket

Includes: Capture Antibody Concentrate

Description

Description

The Human Immunoglobulin M (IgM) Uncoated ELISA Kit contains pre-matched antibody pairs, plates and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IgM. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells.
Specifications

Specifications

15.6-1,000 ng/mL
HRP
Plasma, Serum, Supernatant
ELISA
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22; -76; -86)
10 x 96 Tests
Plasma, 10 μL; Serum, 10 μL; Supernatant, 10 μL
Immunoglobulin M
1 hr. 45 min.
15.6 ng/mL
ELISA Kit
Human
Colorimetric Microplate Reader
HRP
RUO
2°C to 8°C
Human
24 hr. 30 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only