Learn More
STEMCELL Technologies Human IgM ELISA Antibody Pair Kit
Description
- The Human Immunoglobulin M (IgM) ELISA Antibody Pair Kit is intended for those who want the flexibility of setting up their own ELISA assay
- The kit includes capture and detection antibodies and an IgM standard
- It is designed for the quantitative detection and measurement of human IgM in biological fluids such as serum, plasma, and cell culture supernatants
- IgM is predominantly found in lymph fluid and in blood where it acts as a neutralizing agent in the early stages of infection
- Secreted IgM exists in a pentameric form whereas monomeric IgM is found on the surface of B cells
- IgM constitutes approximately 10% of all serum immunoglobulins
- NOTE: This kit includes sufficient reagents for 6 x 96-well ELISA plates (Catalog #38019)
- ELISA plates and pNPP Substrate (Catalog #01917) are required for use with the Human IgM ELISA Antibody Pair Kit and are available for purchase separately
Specifications
Specifications
| Description | B Cells; Hybridomas |
| Quantity | 1 Kit for 6 Plates |