missing translation for 'onlineSavingsMsg'
Learn More

Human IgM Antibody (ICL-931), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18418623 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418623 0.2 mL 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418623 Supplier Novus Biologicals Supplier No. NBP142268AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Human IgM Monoclonal antibody specifically detects Human IgM in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Radioimmune Assays, Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgM
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Radioimmune Assays (RIA)
Classification Monoclonal
Clone ICL-931
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Radioimmunoassay, Dot Blot
Formulation 50mM Sodium Borate
Gene Alias AGM1, MU, VH
Host Species Mouse
Immunogen Purified human IgM
Purification Method Ion exchange chromatography
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.