missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IgM Antibody (3H10G5F2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51525-0.025mg
This item is not returnable.
View return policy
Description
Human IgM Monoclonal specifically detects Human IgM in Human samples. It is validated for ELISA.
Specifications
| Human IgM | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA 1: 10000 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 |
| ELISA | |
| 3H10G5F2 | |
| Unconjugated | |
| AGM1, MU, VH | |
| ChromPure Human IgM (myeloma), whole molecule. | |
| 0.025 mg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction