missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IGHMBP2 (aa 583-658) Control Fragment Recombinant Protein

Product Code. 30197286
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30197286 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30197286 Supplier Invitrogen™ Supplier No. RP107037

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (%), Rat (%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66357 (PA5-66357. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

5' to 3' helicase that unwinds RNA and DNA duplices in an ATP-dependent reaction. Acts as a transcription regulator. Required for the transcriptional activation of the flounder liver-type antifreeze protein gene. Exhibits strong binding specificity to the enhancer element B of the flounder antifreeze protein gene intron. Binds to the insulin II gene RIPE3B enhancer region. May be involved in translation. DNA-binding protein specific to 5'-phosphorylated single-stranded guanine-rich sequence related to the immunoglobulin mu chain switch region. Preferentially binds to the 5'-GGGCT-3' motif. Interacts with tRNA-Tyr. Stimulates the transcription of the human neurotropic virus JCV.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P38935
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 3508
Name Human IGHMBP2 (aa 583-658) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AEP; antifreeze enhancer-binding protein; antifreeze enhancer-binding protein ortholog; ATP-dependent helicase IGHMBP2; cardiac transcription factor 1; CATF1; CMT2S; DNA-binding protein SMUBP-2; GF-1; Glial factor 1; HCSA; HMN6; Ighmbp2; immunoglobulin mu binding protein 2; immunoglobulin mu DNA binding protein 2; immunoglobulin mu-binding protein 2; immunoglobulin S mu binding protein 2; neuromuscular degeneration; nmd; p110 subunit; RIPE3b1; sma; SMARD1; Smbp2; Smbp-2; SMUBP2; ZFAND7; zinc finger, AN1-type domain 7
Common Name IGHMBP2
Gene Symbol IGHMBP2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NRKGEVGFLAEDRRINVAVTRARRHVAVICDSRTVNNHAFLKTLVEYFTQHGEVRTAFEYLDDIVPENYSHENSQG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.