missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IgG4 Recombinant Rabbit Monoclonal Antibody (K01_2Q16)

Product Code. 30826613
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826613 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30826613 Supplier Invitrogen™ Supplier No. MA565707

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: DKRVESKYGP PCPSC.

Human IgG4, also known as human immunoglobulin G4, is a subclass of human IgG antibodies that is produced by the immune system in response to antigens. It is the least abundant subclass of IgG in the blood and plays a minor role in defense against pathogens. IgG4 works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG4 is also involved in other immune responses, such as immune tolerance and suppression of inflammation. Human IgG4 can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2Q16
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene IGHG4
Gene Accession No. P01861
Gene Alias IGHG4; Immunoglobulin G; Immunoglobulin G4; immunoglobulin heavy constant gamma 4 (G4m marker); ImmunoglobulinG
Gene Symbols IGHG4
Host Species Rabbit
Immunogen A synthesized peptide derived from human IgG4 (aa 95-109).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3503
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.