missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IgG4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£178.00 - £454.00
Specifications
| Antigen | HumanIgG4 |
|---|---|
| Clone | 985547 |
| Dilution | Western Blot 2μg/mL, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18121531
![]() |
R&D Systems
MAB9895-SP |
25 μg |
£178.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18113131
![]() |
R&D Systems
MAB9895-100 |
100 μg |
£454.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Human IgG4 Monoclonal specifically detects Human IgG4 in Human samples. It is validated for Western Blot, ELISA.Specifications
| HumanIgG4 | |
| Western Blot 2μg/mL, ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Ig gamma-4 chain C region, immunoglobulin heavy constant gamma 4 (G4m marker), MGC117419 | |
| Synthetic peptide containing human IgG 4 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human IgG 4 in direct ELISAs. |
| 985547 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| 3503 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title