missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IgG1 (103 Cys/Ser) Fc Chimera Recombinant Protein

Product Code. 17164223
Click to view available options
Quantity:
5 μg
5 x 5 μg
5 x 50 μg
50 μg
Unit Size:
50µg
5µg
Pack of 5
This item is not returnable. View return policy

Product Code. 17164223

Brand: Invitrogen™ A42558

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Immunoglobulin G (IgG) is synthesized and secreted by plasma B cells, and constitutes 75% of serum immunoglobulins in humans. IgG antibodies protect the body against the pathogens by agglutination and immobilization, complement activation, toxin neutralization, as well as the antibody-dependent cell-mediated cytotoxicity (ADCC). Recombinant IgG Fc is suggested to represent a potential anti-inflammatory drug for treatment of human autoimmune diseases.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Control, ELISA, Functional Assay, Immunohistochemistry, Western Blot
Formulation PBS with 0.02% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4
Name Human IgG1 Fc
Quantity 5 x 5 μg
Source HEK293 cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias IgG; IgG Fc; IgG1; IgG1 heavy chain; Immunoglobulin gamma-1 heavy chain
Specific Reactivity Human
Conjugate Unconjugated
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥ 95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.