missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IgG1 (103 Cys/Ser) Fc Chimera Recombinant Protein
Click to view available options
Quantity:
5 μg
5 x 5 μg
5 x 50 μg
50 μg
Unit Size:
50µg
5µg
Pack of 5
Description
Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Constant region of immunoglobulin heavy chains. Immunoglobulins, also known as antibodies, are membrane-bound or secreted glycoproteins produced by B lymphocytes. In the recognition phase of humoral immunity, the membrane-bound immunoglobulins serve as receptors which, upon binding of a specific antigen, trigger the clonal expansion and differentiation of B lymphocytes into immunoglobulins-secreting plasma cells. Secreted immunoglobulins mediate the effector phase of humoral immunity, which results in the elimination of bound antigens (PubMed:22158414, PubMed:20176268). The antigen binding site is formed by the variable domain of one heavy chain, together with that of its associated light chain. Thus, each immunoglobulin has two antigen binding sites with remarkable affinity for a particular antigen. The variable domains are assembled by a process called V-(D)-J rearrangement and can then be subjected to somatic hypermutations which, after exposure to antigen and selection, allow affinity maturation for a particular antigen (PubMed:17576170, PubMed:20176268). [UniProt].
Specifications
Specifications
| Accession Number | P01857 |
| For Use With (Application) | Control, ELISA Standard, Binding Assay, Western Blot |
| Formulation | PBS with 0.01% Tween 80, 5% mannitol, 5% trehalose and no preservative; pH 7.4 |
| Gene ID (Entrez) | 3500 |
| Molecular Weight (g/mol) | 26 kDa |
| Name | Human IgG1 (103 Cys/Ser) Fc Chimera |
| pH Range | 7.4 |
| Purification Method | Purified |
| Quantity | 5 μg |
| Source | HEK293 cells |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction