missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IgG1 Chimeric Isotype Control (MOPC-21), Invitrogen™
Human Recombinant Monoclonal Isotype Control
Brand: Invitrogen MA547824
Additional Details : Weight : 0.01000kg
Description
Specificity: An antibody of unknown specificity that maybe used as a negative control.
Human IgG1, also known as human immunoglobulin G1, is a subclass of human IgG antibodies that is produced by the immune system in response to antigens. It is the most abundant subclass of IgG in the blood and plays a key role in defense against pathogens. IgG1 works by binding to specific antigens and promoting their clearance from the body through various mechanisms, including phagocytosis and complement activation. IgG1 is also involved in other immune responses, such as antibody-dependent cellular cytotoxicity and neutralization of toxins. Human IgG1 can be used in research to study the immune response to pathogens and to develop diagnostic tests and therapeutic interventions for infectious and autoimmune diseases.Specifications
Human IgG1 Chimeric | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
O35516, Q04721 | |
Human | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |
Control | |
MOPC-21 | |
Unconjugated | |
NOTCH2 | |
IGHG1; immunoglobulin heavy constant gamma 1 (G1m marker) | |
Proprietary sequence. | |
200 μg | |
18129, 4853 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 κ |