missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IgG Fc Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
£187.00 - £1775.00
Specifications
| Antigen | IgG |
|---|---|
| Clone | 97924 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18791127
![]() |
R&D Systems
MAB110-SP |
25 μg |
£187.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18718378
![]() |
R&D Systems
MAB110-100 |
100 μg |
£448.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18758408
![]() |
R&D Systems
MAB110-500 |
500 μg |
£1775.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgG Monoclonal specifically detects IgG in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| IgG | |
| LYOPH | |
| Western Blot, Flow Cytometry, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 3500 | |
| Mouse myeloma cell line NS0-derived recombinant human IgG 1 Fc region | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects chimeric proteins containing a human IgG Fc region in direct ELISAs and Western blots. |
| 97924 | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| Immunoglobulin G, ImmunoglobulinG | |
| IGHG1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title