missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Mouse IgG ELISA Kit (Chemiluminescence)

Product Code. 18391394 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18391394 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18391394 Supplier Novus Biologicals™ Supplier No. NBP300441

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human IgG ELISA Kit (Chemiluminescence) measures IgG in Serum, plasma, cell supernatant, and other biological fluids.

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.31 to 20 ng/mL
Assay Sensitivity 0.19 ng/mL
Product Type ELISA Kit (Chemiluminescence)
Sample Type Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
Specificity This kit recognizes Mouse IgG in samples. No significant cross-reactivity or interference between Mouse IgG and analogues was observed.
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 3500
Gene Symbol IGHG1
Quantity 1 Kit
Research Discipline Adaptive Immunity, Epitope Tags, Immunology
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym Immunoglobulin G, ImmunoglobulinG
Target IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.