missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IGFBP-rp1/IGFBP-7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£143.00 - £420.00
Specifications
| Antigen | IGFBP-rp1/IGFBP-7 |
|---|---|
| Clone | 192535 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18782507
![]() |
R&D Systems
MAB13342-SP |
25 μg |
£143.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18704437
![]() |
R&D Systems
MAB13342-100 |
100 μg |
£420.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGFBP-rp1/IGFBP-7 Monoclonal specifically detects IGFBP-rp1/IGFBP-7 in Human samples. It is validated for ELISA.Specifications
| IGFBP-rp1/IGFBP-7 | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| AAA16187 | |
| 3490 | |
| IgG2a | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 192535 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| angiomodulin, FSTL2, IBP-7, IGFBP7, IGFBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBPrp1, IGFBP-rP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, Mac25, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSF, PSFAGM, TAF, Tumor-derived adhesion factor | |
| Mouse myeloma cell line NS0-derived recombinant human IGFBP-rp1/IGFBP-7, Asp30-Arg277, Accession # AAA16187 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IGFBP-rp1/IGFBP-7 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title