missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IGFBP-rp1/IGFBP-7 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
£168.00 - £1755.00
Specifications
| Antigen | IGFBP-rp1/IGFBP-7 |
|---|---|
| Clone | 192520 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, ELISA |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18775519
![]() |
R&D Systems
MAB1334-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18763027
![]() |
R&D Systems
MAB1334-100 |
100 μg |
£441.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18745558
![]() |
R&D Systems
MAB1334-500 |
500 μg |
£1755.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IGFBP-rp1/IGFBP-7 Monoclonal specifically detects IGFBP-rp1/IGFBP-7 in Human samples. It is validated for Western Blot, ELISA.Specifications
| IGFBP-rp1/IGFBP-7 | |
| LYOPH | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| angiomodulin, FSTL2, IBP-7, IGFBP7, IGFBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBPrp1, IGFBP-rP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, Mac25, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSF, PSFAGM, TAF, Tumor-derived adhesion factor | |
| IGFBP7 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 192520 | |
| Western Blot 1 ug/mL, ELISA | |
| Monoclonal | |
| Mouse | |
| Human | |
| AAA16187 | |
| 3490 | |
| Mouse myeloma cell line NS0-derived recombinant human IGFBP-rp1/IGFBP-7 Asp30-Arg277 Accession # AAA16187 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IGFBP-rp1/IGFBP-7 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human IGFBP-1, -2, -3, -4, -5, or -6 is observed. |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto