missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IGFBP-rp1/IGFBP-7 Antibody, R&D Systems™

Goat Monoclonal Antibody
Brand: R&D Systems MAB13341-100
This item is not returnable.
View return policy
Description
IGFBP-rp1/IGFBP-7 Monoclonal specifically detects IGFBP-rp1/IGFBP-7 in Human samples. It is validated for Immunohistochemistry.
Specifications
| IGFBP-rp1/IGFBP-7 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| angiomodulin, FSTL2, IBP-7, IGFBP7, IGFBP-7, IGFBP-7PGI2-stimulating factor, IGFBP-7v, IGFBPrp1, IGFBP-rP1, insulin-like growth factor binding protein 7, insulin-like growth factor-binding protein 7, Mac25, MAC25 protein, MAC25IGF-binding protein 7, Prostacyclin-stimulating factor, PSF, PSFAGM, TAF, Tumor-derived adhesion factor | |
| Mouse myeloma cell line NS0-derived recombinant human IGFBP-rp1 Arg98-Arg277 Accession # AAA16187 | |
| 100 μg | |
| Primary | |
| Detects human IGFBP-rp1 in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 40012B | |
| Immunohistochemistry 1-25 ug/mL | |
| AAA16187 | |
| Goat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 3490 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction