missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IGFBP-L1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF3877
This item is not returnable.
View return policy
Description
IGFBP-L1 Polyclonal specifically detects IGFBP-L1 in Human samples. It is validated for Western Blot.
Specifications
| IGFBP-L1 | |
| Polyclonal | |
| Biotin | |
| Q8WX77 | |
| IGFBPL1 | |
| Mouse myeloma cell line NS0-derived recombinant human IGFBP-L1 Ser25-Met278 Accession # Q8WX77 | |
| 50 μg | |
| Primary | |
| Detects human IGFBP-L1 in Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse (rm) IGFBP-L1 is observed and less than 1% cross-reactivity with recombinant human (rh) IGFBP-2, -3, -4, -5, -6, -7, rmIGFBP-1, and rmIGFBP-rP10 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| bA113O24.1, IGFBPL1, IGFBPL1 insulin-like growth factor binding protein-like 1, IGFBP-RP4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 347252 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction